missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Legionella Latex Test

Rapidly confirm predominant Legionella serogroups grown on selective media by latex agglutination.

Brand:  Thermo Scientific™ DR0803M

239.95 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11463949

  • £251.00 / 50 tests

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Rapidly differentiate and confirm the identity of predominant Legionella serogroups grown on selective media by latex agglutination using Thermo Scientific™ Legionella Latex Test.

Use this latex agglutination test for the identification of predominant Legionella species grown on plated media from patients with suspected Legionellosis or from environmental sources. The test allows separate identification of Legionella pneumophila serogroup 1 and serogroups 2-14 and detection of seven other Legionella species which have been implicated in human disease.

Blue latex particles sensitised with antibody agglutinate in the presence of specific Legionella cell wall antigens to form visible clumps.

The following products are available for use with the Legionella Latex Test and are sold separately:

Legionella Pneumophila Serogroup 1 Test Reagent (Part No. DR0801M)

Legionella Pneumophila Serogroup 2-14 Test Reagent (Part No. DR0802M)

Legionella Species Test Reagent (Part No. DR0800M)—Legionella longbeachae 1 & 2, Legionella bozemanii 1 & 2, Legionella dumoffii, Legionella gormanii, Legionella jordanis, Legionella micdadei and Legionella anisa

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2°C to 8°C
Legionella
Legionella Latex Test Kit
Legionella Species Test Reagent
Environmental Source
50 Tests/Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6191496 Certificate TypeCertificate of Analysis Date03/02/2025 Catalog NumberDR0803M
Lot Number6191495 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog NumberDR0803M
Lot Number6170684 Certificate TypeCertificate of Analysis Date05/12/2024 Catalog NumberDR0803M
Lot Number6143613 Certificate TypeCertificate of Analysis Date04/11/2024 Catalog NumberDR0803M
Lot Number6156364 Certificate TypeCertificate of Analysis Date01/11/2024 Catalog NumberDR0803M
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Legionella Latex Test >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.