missing translation for 'onlineSavingsMsg'
Learn More
uvex Lens Cleaning Tissues Product Code.: 16430351
In Stock
to see stock.
Add to basket

uvex Lens Cleaning Tissues

Product Code. 16430351 Shop All Uvex Products
Lens Cleaning Tissues
Click to view available options
Lens Cleaning Tissues

Product Code. 16430351

Brand: uvex 9971000

In Stock
to see stock.
Edge
Add to basket
Add to basket

Tissues are suitable for all lenses to ensure that users can see clearly at all times.

  • Suitable for use with all Uvex spectacles and goggles
TRUSTED_SUSTAINABILITY

Specifications

Color White
Product Type Lens Cleaning Tissue
For Use With (Equipment) Uvex™ Safety Spectacles
Disposable Reusable
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131

    Expires2025-06-08

    New Offer
    Shop Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
uvex Lens Cleaning Tissues >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.