Learn More
Leupeptin, MP Biomedicals™
Inhibits trypsin-like proteases and some cysteine proteases. MP Biomedicals' Leupeptin is a modified tripeptide reversible protease inhibitor. Little to no inhibition is seen against pepsin, cathepsins A and D, and alpha-chymotrypsin.
Brand:  MP Biomedicals 0215155380
Code : NEW
Additional Details : CAS Number : 103476-89-7 Weight : 0.00010kg
 View more versions of this product
Product Code. 11458191
  • £736.70 / 100mg
Estimated Shipment: 06-08-2024
to see stock.
Description
Description
When adjusted for molarity, all three salt forms are equally effective; however, the hydrochloride salt is usually less invasive in biological settings.
Inhibits trypsin-like proteases and some cysteine proteases including: endoproteinase Lys-C, kallikrein, papain, thrombin, cathepsin B, cathepsins H and L, trypsin, calpain, trypsin8 and plasmin.
Specifications
Specifications
103476-89-7
White Powder
100mg
Store at −20°C, desiccate, protect from light.
427.57