Learn More
Leupeptin, MP Biomedicals™
Inhibits trypsin-like proteases and some cysteine proteases. MP Biomedicals' Leupeptin is a modified tripeptide reversible protease inhibitor. Little to no inhibition is seen against pepsin, cathepsins A and D, and alpha-chymotrypsin.
£79.71 - £736.70
Especificaciones
CAS | 103476-89-7 |
---|---|
Content And Storage | Store at −20°C, desiccate, protect from light. |
Format | White Powder |
Molecular Weight (g/mol) | 427.57 |
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
11438191
|
MP Biomedicals
0215155305 |
5mg |
£79.71
5mg |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11448191
|
MP Biomedicals
0215155325 |
25mg |
£261.82
25mg |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11458191
|
MP Biomedicals
0215155380 |
100mg |
£736.70
100mg |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descripción
When adjusted for molarity, all three salt forms are equally effective; however, the hydrochloride salt is usually less invasive in biological settings.
Inhibits trypsin-like proteases and some cysteine proteases including: endoproteinase Lys-C, kallikrein, papain, thrombin, cathepsin B, cathepsins H and L, trypsin, calpain, trypsin8 and plasmin.
Especificaciones
103476-89-7 | |
Store at −20°C, desiccate, protect from light. | |
White Powder | |
427.57 |