Learn More
Elma™ Level Sets
Modular basket system for the Elmasonic P, Elmasonic Select, and EASY.
Brand:  Elma™ 1125226
 View more versions of this product
Product Code. 17841347
  • £12.95 / 1 set
Estimated Shipment: 14-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stainless steel with plastic handles (10 to 120 and 180 to 300) or Rilsan coating (150)
  • Ergonomic handles
  • Autoclavable
  • Bayonet holes in the bottom for mounting holders (e.g. Elma Pin, silicone holder bottom comb)
  • Square openings in the side walls for mounting silicone holders
Specifications
Specifications
Level Set
Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY
M