Learn More
Elma™ Level Sets
Modular basket system for the Elmasonic P, Elmasonic Select, and EASY.
£12.95 - £30.15
Specifications
Type | Level Set |
---|---|
For Use With (Equipment) | Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY |
Product Code | Brand | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | |||||
17891327
|
Elma™
1125225 |
S |
£12.95
1 set |
Estimated Shipment: 14-11-2024
Log in to see stock. |
|||||
17841347
|
Elma™
1125226 |
M |
£12.95
1 set |
Estimated Shipment: 14-11-2024
Log in to see stock. |
|||||
17811337
|
Elma™
1125227 |
L |
£25.85
Each |
Estimated Shipment: 14-11-2024
Log in to see stock. |
|||||
17831357
|
Elma™
1125228 |
XL |
£30.15
Each |
Estimated Shipment: 14-11-2024
Log in to see stock. |
|||||
Description
- Stainless steel with plastic handles (10 to 120 and 180 to 300) or Rilsan coating (150)
- Ergonomic handles
- Autoclavable
- Bayonet holes in the bottom for mounting holders (e.g. Elma Pin, silicone holder bottom comb)
- Square openings in the side walls for mounting silicone holders
Specifications
Level Set | |
Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY |