Learn More
Levofloxacin, MedChemExpress
MedChemExpress Levofloxacin ((-)-Ofloxacin) is an orally active antibiotic and is active against both Gram-positive and Gram-negative bacteria. Levofloxacin inhibits the DNA gyrase and topoisomerase IV. Levofloxacin can be used for chronic periodontitis, airway inflammation and BK Viremia research. Levofloxacin shows anti-orthopoxvirus activity.
Brand: Medchem Express HY-B0330
Description
Levofloxacin ((-)-Ofloxacin) is an orally active antibiotic and is active against both Gram-positive and Gram-negative bacteria. Levofloxacin inhibits the DNA gyrase and topoisomerase IV. Levofloxacin can be used for chronic periodontitis, airway inflammation and BK Viremia research. Levofloxacin shows anti-orthopoxvirus activity.Specifications
Levofloxacin | |
100986-85-4 | |
5 g | |
C18H20FN3O4 | |
(-)-Ofloxacin | |
O=C(C(C1=O)=CN2[C@@H](C)COC3=C(N4CCN(C)CC4)C(F)=CC1=C23)O | |
361.37 | |
Research |
Powder | |
Off-White | |
Research | |
COVID-19-anti-virus | |
H2O : 50 mg/mL (138.36 mM; Need ultrasonic) ∣DMSO : 10 mg/mL (27.67 mM; Need ultrasonic) | |
361.37 | |
98.76% |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only