Learn More
Levofloxacin, MedChemExpress
MedChemExpress Levofloxacin ((-)-Ofloxacin) is an orally active antibiotic and is active against both Gram-positive and Gram-negative bacteria. Levofloxacin inhibits the DNA gyrase and topoisomerase IV. Levofloxacin can be used for chronic periodontitis, airway inflammation and BK Viremia research. Levofloxacin shows anti-orthopoxvirus activity.
£75.02 - £105.27
Specifications
Chemical Name or Material | Levofloxacin |
---|---|
Physical Form | Powder |
CAS | 100986-85-4 |
Color | Off-White |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16488236
|
Medchem Express
HY-B0330 |
100 mg |
£75.02
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16480878
|
Medchem Express
HY-B0330 |
5 g |
£105.27
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Levofloxacin ((-)-Ofloxacin) is an orally active antibiotic and is active against both Gram-positive and Gram-negative bacteria. Levofloxacin inhibits the DNA gyrase and topoisomerase IV. Levofloxacin can be used for chronic periodontitis, airway inflammation and BK Viremia research. Levofloxacin shows anti-orthopoxvirus activity.Specifications
Levofloxacin | |
100986-85-4 | |
Research | |
COVID-19-anti-virus | |
H2O : 50 mg/mL (138.36 mM; Need ultrasonic) ∣DMSO : 10 mg/mL (27.67 mM; Need ultrasonic) | |
361.37 | |
98.76% |
Powder | |
Off-White | |
C18H20FN3O4 | |
(-)-Ofloxacin | |
O=C(C(C1=O)=CN2[C@@H](C)COC3=C(N4CCN(C)CC4)C(F)=CC1=C23)O | |
361.37 | |
Research |
Safety and Handling
H302∣H317∣H334∣H361∣H362
missing translation for 'shelfLife' : 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Recommended Storage : 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.