Learn More
LHPP Mouse anti-Mouse, Unconjugated, Clone: 2A2A4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66988-1-IG-150UL
This item is not returnable.
View return policy
Description
The LHPP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LHPP. This antibody recognizes mouse, rat, pig antigen. The LHPP antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
LHPP | |
Monoclonal | |
913 μg/mL | |
Mouse | |
Primary | |
Mouse, Rat, Pig |
Western Blot, Immunofluorescence, ELISA | |
2A2A4 | |
Unconjugated | |
150 μL | |
64077 | |
IgG1 |