missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LHPP Mouse anti-Mouse, Unconjugated, Clone: 2A2A4, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
Antigen | LHPP |
---|---|
Clone | 2A2A4 |
Concentration | 913 μg/mL |
Applications | Western Blot, Immunofluorescence, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16829335
|
Proteintech
66988-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16819335
|
Proteintech
66988-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The LHPP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LHPP. This antibody recognizes mouse, rat, pig antigen. The LHPP antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
LHPP | |
913 μg/mL | |
Monoclonal | |
Mouse | |
64077 | |
Primary |
2A2A4 | |
Western Blot, Immunofluorescence, ELISA | |
Unconjugated | |
Mouse, Rat, Pig | |
IgG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title