Learn More
Cytiva Life Sciences rProtein A Protein GraviTrap™
Includes ten ready-to-use columns prepacked with 1ml of a mixture of 50% rProtein A Sepharose Fast Flow and 50% Protein G Sepharose 4 Fast Flow
Brand: Cytiva 28-9852-56
Code : GH
Additional Details : CAS Number : 64-17-5 Weight : 0.62370kg
Description
Prepacked gravity-flow columns provide convenience, save time, and increase the reliability of your results.
- Purification of antibodies in 30 min. allows you to quickly obtain and store your target protein in order to avoid the inherent dangers of protein instability and degradation associated with slow and long purification runs
- High purity and recovery with purity >95% in one step and recovery typically 70% to 80%
- High binding capacity such as Protein A/Protein G GraviTrap approximately 35mg human IgG/column
- Antibodies are simply captured with high specificity on protein A and protein G ligands in gravity-flow columns
- Yield varies from 20 to 50mg of purified antibodies depending on the ligand you use
- Columns are reusable up to five times
- Complete pack size: 10 columns
- Application: Manual purification of monoclonal and polyclonal antibodies
- Includes: 1 pack of 10 × 1mL columns
- Binding capacity: Approx. 35mg human IgG/column
- Bed volume: 1mL
- Material (Column hardware): Polypropylene barrel, Polyethylene frits
Specifications
1 mL | |
rProtein A Protein G GraviTrap contains ten columns with 1 mL 50% rProtein A Sepharose Fast Flow and 50% Protein G Sepharose 4 Fast Flow resins. | |
2°C to 8°C | |
Gravity flow |
1 mL | |
10 Ready-to -use column prepacked with 1mL of a mixture of 50% rProtein A Sepharose fast flow and 50% Protein G Sepharose 4 fast flow, Polypropylene barrel and Polyethylene frits | |
10 Pack | |
Gravity Flow Column |
Safety and Handling
- rProtein A/Protein G GraviTrap
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.