missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Purolite™ Lifetech™ ECR Resins for Enzyme Immobilization, ECR8285
Lifetech™ ECR resins are robust polymeric resins designed for research and industrial immobilization of enzymes. Available for covalent, ionic, adsorption and affinity immobilization.
£167.00 - £473.00
Specifications
Description | White to off-white spherical beads |
---|---|
pH Range | 5 to 9 |
Pore Size | 400 to 600 Å |
Content And Storage | 2°C to 8°C |
Particle Size | 250 to 1000 μm |
Description
- Enzyme immobilization by adsorption, covalent, IEX and affinity
- High mechanical stability
- High porosity
- Easily dried after immobilization
- Suitable for batch and column configuration
- Optimal for use in organic solvent/water free systems
- Compliant with ResAP(2004) 3
- Halal, Kosher and TSE/BSE/GMO free
Specifications
White to off-white spherical beads | |
400 to 600 Å | |
250 to 1000 μm | |
Epoxy | |
Methacrylate (ECR Resin) |
5 to 9 | |
2°C to 8°C | |
70 to 80% | |
Wet |
Safety and Handling
missing translation for 'shelfLife' : Use within 6 months from date of manufacture