missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Purolite™ Lifetech™ Immobilized Enzymes
Click to view available options
Product Line:
CalB immo Plus
CalB immo Plus™ Food Grade
CalB immo™ 8285
CalB immo™ 8806
Quantity:
10 g
50 g
250 g
1 kg
Unit Size:
10g
1kg
250g
50g
Description
- Use in solvent-free or organic solvent systems
- Esterifications (regio- and stereo-selective)
- Transesterfication
- Amidation
- Fats and oils modification
- Hydrolysis of esters
- High mechanical stability
- High enzyme activity
- Organic solvent stability
- Available in large volumes
- Security of supply from Kg to ton scale
- Resin compliant with ResAP(2004) 3
- Halal and TSE/BSE/GMO free
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Particle Size | 300 to 710 μm |
| Moisture | <5% |
| Form | Dry |
| Product Line | CalB immo™ 8806 |
| Quantity | 250 g |
| Target | Enzyme Activity: >10000 PLU/gdry |
| Type | Immobilized Lipase |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction