missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Purolite™ Lifetech™ Immobilized Enzymes
Lipase immobilized on robust polymeric resins and are supplied in dry form. They can be used for esterifications, amidations, transesterifications, chiral separations. Can be used in water, organic phase, bi-phasic systems and oil phase.
Brand: Purolite™ LS02031-94
This item is not returnable.
View return policy
Description
- Use in solvent-free or organic solvent systems
- Esterifications (regio- and stereo-selective)
- Transesterfication
- Amidation
- Fats and oils modification
- Hydrolysis of esters
- High mechanical stability
- High enzyme activity
- Organic solvent stability
- Available in large volumes
- Security of supply from Kg to ton scale
- Resin compliant with ResAP(2004) 3
- Halal and TSE/BSE/GMO free
Specifications
White to slightly yellow spherical beads | |
250 to 1000 μm | |
Dry | |
250 g | |
Immobilized Lipase |
2°C to 8°C | |
<5% | |
CalB immo™ 8285 | |
Enzyme Activity: >10000 PLU/gdry |