missing translation for 'onlineSavingsMsg'
Learn More

Purolite™ Lifetech™ Immobilized Enzymes

Product Code. 17099383
Click to view available options
Product Line:
CalB immo Plus
CalB immo Plus™ Food Grade
CalB immo™ 8285
CalB immo™ 8806
Quantity:
10 g
50 g
250 g
1 kg
Unit Size:
10g
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 17099383

Brand: Purolite™ LS02009192

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Lipase immobilized on robust polymeric resins and are supplied in dry form. They can be used for esterifications, amidations, transesterifications, chiral separations. Can be used in water, organic phase, bi-phasic systems and oil phase.

  • Use in solvent-free or organic solvent systems
  • Esterifications (regio- and stereo-selective)
  • Transesterfication
  • Amidation
  • Fats and oils modification
  • Hydrolysis of esters
  • High mechanical stability
  • High enzyme activity
  • Organic solvent stability
  • Available in large volumes
  • Security of supply from Kg to ton scale
  • Resin compliant with ResAP(2004) 3
  • Halal and TSE/BSE/GMO free

Specifications

Content And Storage 2°C to 8°C
Particle Size 300 to 710 μm
Moisture <5%
Form Dry
Product Line CalB immo Plus
Quantity 50 g
Target Enzyme Activity: >9000 PLU/gdry
Type Immobilized Lipase
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.