missing translation for 'onlineSavingsMsg'
Learn More

3M™ Safety-Walk™ Light Duty Cushion Matting 3270E

Spring coiled, vinyl matting that compresses uniformly and continually. Helps provide greater comfort and less fatigue.

Brand:  3M™ Safety-Walk™ 7100116725

Additional Details : Weight : 14.38300kg

 View more versions of this product

Product Code. 17158740

  • £740.00 / Each
Estimated Shipment: 19-08-2024
to see stock.
Add to basket

Description

Description

  • Coiled cushion matting compresses evenly across the bottom of the shoe to reduce leg and back fatigue
  • Textured top improves traction for safer footing
  • Suitable for dry areas such as assembly lines, packing areas, and security posts
  • Supplied with edging to allow wheeled traffic
  • Special vinyl coils to withstand most industrial and cleaning chemicals
Specifications

Specifications

10 ft.
3 ft.
1 Ea.
3.1 m
Black
Cushion Mat
914 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers