Learn More
Lauda™ Light Yellow Heat Transfer Liquid
Brand: Lauda™ LZB 209
Additional Details : Weight : 10.80000kg
Specifications
–30 to 90°C | |
-30°C to +90°C | |
Kryo 30 | |
4mm2/sec. |
10 L | |
Bath Filler | |
10 L |
Safety and Handling
- Bath liquid KRYO 30 (1 can à 10 litres) .
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands and face thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethylene glycol