missing translation for 'onlineSavingsMsg'
Learn More

Lincomycin, hydrochloride, ≥82%, MP Biomedicals™

Lincomycin, Hydrochloride

Marke:  MP Biomedicals 0215894805


 Lernen Sie weitere Varianten dieses

Artikelnummer. 11454212

  • £147.00 / 5g

Please to purchase this item. Need a web account? Register with us today!

Entdecken Sie weitere Sonderangebote
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Beschreibung

Beschreibung

Lincomycin hydrochloride is used to study the inhibition of protein synthesis, bacterial antibiotic resistance, and protein and surfactant binding.
TRUSTED_SUSTAINABILITY
Spezifikation

Spezifikation

Lincomycin hydrochloride
Powder
White
≥82%
Lincocin hydrochloride, Methyl 6,8-dideoxy-6-(1-methyl-4-propyl-2-pyrrolidinecarboxamido)-1-thio-D-erythro-α-D-galactooctopyranoside hydrochloride, U-10149A
≥82%
Lincomycin Hydrochloride
859-18-7
5 g
C18H34N2O6S·HCl
442.996
Produktvorschläge

Produktvorschläge

Videos
Sicherheit und Handhabung

Sicherheit und Handhabung

Produktidentifikator
  • LINCOMYCIN HYDROCHLORIDE

Signalwort
  • Warning

Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Gefahrenhinweis
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Sicherheitshinweis
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Ergänzende Informationen
  • MIXTURE LIST-Contains : Lincomycin Hydrochloride

Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.