missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lincomycin, hydrochloride, ≥82%, MP Biomedicals™
Lincomycin, Hydrochloride
£53.00 - £146.28
Specifications
Type | Lincomycin Hydrochloride |
---|---|
Physical Form | Powder |
CAS | 859-18-7 |
Assay Percent Range | ≥82% |
Molecular Formula | C18H34N2O6S·HCl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17680877
|
MP Biomedicals
0215894801 |
1 g |
£53.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11454212
|
MP Biomedicals
0215894805 |
5 g |
£146.28
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lincomycin hydrochloride is used to study the inhibition of protein synthesis, bacterial antibiotic resistance, and protein and surfactant binding.Specifications
Lincomycin Hydrochloride | |
859-18-7 | |
C18H34N2O6S·HCl | |
442.996 | |
White |
Powder | |
≥82% | |
Lincocin hydrochloride, Methyl 6,8-dideoxy-6-(1-methyl-4-propyl-2-pyrrolidinecarboxamido)-1-thio-D-erythro-α-D-galactooctopyranoside hydrochloride, U-10149A | |
≥82% |
Safety and Handling
missing translation for 'storageNote' : Room Temperature
Recommended Storage : Store at 4°C.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.