missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Linerless Pipe Markers - Standard Legends

European Standard Pipemarkers reinforce safety and aid maintenance.

Brand:  Brady™ N006988

Code : NEW

Additional Details : Weight : 0.10600kg

 View more versions of this product

Product Code. 15349015

  • £102.00 / 1 roll
Estimated Shipment: 23-07-2024
to see stock.
Add to basket

Description

Description

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.
Specifications

Specifications

European Regulations 1271/2008, 790/2009 and 286/2001
Permanent Acrylic
Roll
Pipe Marker
Pipe and valve marking
30 Markers/Roll
-40°F to +284°F
0.0047 in.
33 m x 250 mm
33 m
250 mm
26 mm
Individual linerless Pipe Markers on a Roll with die-cut arrowheads, without pictograms - Acids and Alkalis - Acid
White on Purple
Polyester
Acid
English
30 Pk.
-40°C to +140°C
0.119 mm
108.3 ft.
9.84 in.
1.02 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers