missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Linerless Pipe Markers - Standard Legends
European Standard Pipemarkers reinforce safety and aid maintenance.
£102.00 - £122.00
Specifications
Certifications/Compliance | European Regulations 1271/2008, 790/2009 and 286/2001 |
---|---|
Description | Individual linerless Pipe Markers on a Roll with die-cut arrowheads, without pictograms - Acids and Alkalis - Acid |
Adhesive Type | Permanent Acrylic |
Color | White on Purple |
Format | Roll |
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
15349015
|
Brady™
N006988 |
26 mm |
£102.00
1 roll |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
15359015
|
Brady™
N006989 |
37 mm |
£122.00
1 roll |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
Description
- Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
- Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, saves you having to identify the appropriate symbol.
- Markers are printed with a contrasting border in the same color as the text for improved visibility.
- Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.
Specifications
European Regulations 1271/2008, 790/2009 and 286/2001 | |
Permanent Acrylic | |
Roll | |
Pipe Marker | |
Pipe and valve marking | |
30 Markers/Roll | |
-40°F to +284°F | |
0.0047 in. | |
108.3 ft. |
Individual linerless Pipe Markers on a Roll with die-cut arrowheads, without pictograms - Acids and Alkalis - Acid | |
White on Purple | |
Polyester | |
Acid | |
English | |
30 Pk. | |
-40°C to +140°C | |
0.119 mm | |
33 m |