missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Linoleic acid, 99%
£40.80 - £274.37
Chemical Identifiers
CAS | 60-33-3 |
---|---|
Molecular Formula | C18H32O2 |
Molecular Weight (g/mol) | 280.45 |
InChI Key | OYHQOLUKZRVURQ-HZJYTTRNSA-N |
Synonym | linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid |
PubChem CID | 5280450 |
ChEBI | CHEBI:17351 |
IUPAC Name | (9Z,12Z)-octadeca-9,12-dienoic acid |
SMILES | CCCCCC=CCC=CCCCCCCCC(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10687172
|
Thermo Scientific Chemicals
215040050 |
5 mL | Glass bottle |
£40.80
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10521851
|
Thermo Scientific Chemicals
215040250 |
25 mL | Glass bottle |
£109.55
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11934961
|
Thermo Scientific Chemicals
215041000 |
100 mL | Glass bottle |
£274.37
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
60-33-3 | |
280.45 | |
linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid | |
CHEBI:17351 | |
CCCCCC=CCC=CCCCCCCCC(=O)O |
C18H32O2 | |
OYHQOLUKZRVURQ-HZJYTTRNSA-N | |
5280450 | |
(9Z,12Z)-octadeca-9,12-dienoic acid |
Specifications
60-33-3 | |
Colorless to Yellow | |
229.0°C to 230.0°C (16.0 mmHg) | |
Authentic | |
C18H32O2 | |
1.4687 to 1.4707 | |
15,556 | |
linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid | |
OYHQOLUKZRVURQ-HZJYTTRNSA-N | |
(9Z,12Z)-octadeca-9,12-dienoic acid | |
5280450 | |
280.45 | |
Liquid |
-5.0°C | |
0.9000g/mL | |
>112°C | |
98.5% min. (GC) | |
CH3(CH2)4CH=CHCH2CH=CH(CH2)7CO2H | |
02,IV,1754 | |
0.9 | |
Solubility in water: insoluble. Other solubilities: freely soluble in diethyl ether,miscible with dimethylformamide,soluble in ethanol,petroleum ether | |
CCCCCC=CCC=CCCCCCCCC(=O)O | |
280.45 | |
CHEBI:17351 | |
99% | |
Linoleic acid |
Safety and Handling
Warning
missing translation for 'einecsNumber' : 200-470-9
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title