missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lipofectamine™ 3000 Transfection Reagent
GREENER_CHOICE

Lipofectamine™ 3000 Transfection Reagent leverages our most advanced lipid nanoparticle technology to provide superior transfection performance with improved application outcomes and reproducible results.

£91.25 - £8010.00

Specifications

Classification Animal Origin-Free
Cell Type Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample Type Plasmid DNA, Synthetic siRNA, RNAi Plasmids (shRNA, miR)
Transfection Technique Lipid-based Transfection
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15282465
GREENER_CHOICE
View Documents
Invitrogen™
L3000001
0.1 mL
£91.25
0.10mL
Please to purchase this item. Need a web account? Register with us today!
15292465
GREENER_CHOICE
View Documents
Invitrogen™
L3000008
0.75 mL
£556.00
0.75mL
Please to purchase this item. Need a web account? Register with us today!
15212475
GREENER_CHOICE
View Documents
Invitrogen™
L3000015
1.5 mL
£952.00
1.50mL
Please to purchase this item. Need a web account? Register with us today!
15222475
GREENER_CHOICE
View Documents
Invitrogen™
L3000075
5 x 1.5 mL
£4405.00
7.50mL
Please to purchase this item. Need a web account? Register with us today!
15232475
GREENER_CHOICE
View Documents
Invitrogen™
L3000150
15 mL
£8010.00
15mL
Please to purchase this item. Need a web account? Register with us today!
Description
Show More
Specifications

Specifications

Animal Origin-Free
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Lipid-based Transfection
Sustainable packaging
Transfection Reagent
Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Plasmid DNA, Synthetic siRNA, RNAi Plasmids (shRNA, miR)
Transfection
Lipofectamine™
Yes
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Lipofectamine™ 3000 Transfection Reagent

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.