Learn More
Labconco™ Liquid Trap Kit
For use with CentriVap™ Vacuum Concentrators and RapidVap™ Vacuum Evaporation Systems.
Brand:  Labconco™ 7970500
Product Code. 15363476
  • £496.00 / Each
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Liquid Trap prevents liquid from entering the pump
  • Includes borosilicate glass container that holds 240 milliliters of liquid, epoxy-coated steel bracket for attachment to the side of a CentriVap Concentrator or RapidVap Vacuum Evaporation System, 18 in. (457mm) of 0.5 in. ID (13mm) vacuum tubing, and two vacuum tubing clamps
Specifications
Specifications
Liquid Trap
CentriVap Concentrators and RapidVap Evaporators
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Labconco™ Liquid Trap Kit >