Learn More
Lithium aluminum hydride, 95%, powder, Thermo Scientific Chemicals
£14.87 - £2084.00
Chemical Identifiers
CAS | 16853-85-3 |
---|---|
Molecular Formula | AlH4Li |
MDL Number | MFCD00011075 |
Synonym | lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride |
PubChem CID | 21226445 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10042470
|
Thermo Scientific Acros
190320100 |
10 g | N/A | ||||||
10665832
|
Thermo Scientific Acros
190320250 |
25 g | N/A | ||||||
10123830
|
Thermo Scientific Acros
190321000 |
100 g | N/A | ||||||
10207003
|
Thermo Scientific Acros
190325000 |
500 g | N/A | ||||||
17332481
|
Thermo Scientific Acros
190320050 |
5 kg | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
16853-85-3 | |
MFCD00011075 | |
21226445 |
AlH4Li | |
lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride |
Specifications
125.0°C | |
Powder | |
AlH4Li | |
MFCD00011075 | |
15, 344 | |
Solubility in water: reacts. Other solubilities: soluble in diethyl ether (39g/100g at 25 °C), thf/, toluene (70:30) (17g/100g at 25 °C), in thf/mtbe (, 25:75) (12g/100g at 25 °C), in thf/toluene (15:85), (5g/100g at 25 °C);, practically insoluble in hyd | |
[Li+].[AlH4-] | |
37.95 | |
37.95 | |
.9g/mL |
16853-85-3 | |
95% | |
LiAlH4 | |
01,581; 02,242; 03,176; 04,291; 05,382; 06,325; 07,196; 08,286; 09,274; 10,236; 11,289; 12,272; 13,61; 14,190; 15,184; 16,133; 17,162 | |
lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride | |
OCZDCIYGECBNKL-UHFFFAOYSA-N | |
lithium(1+) alumanuide | |
21226445 | |
95% | |
Lithium Aluminum hydride |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- Lithium aluminium hydride
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
Precautionary Statement
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.