Learn More
Lithium borohydride, 4M (10 wt.%) solution in THF, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 428888000
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
Lithium borohydride | |
800 mL | |
LiBH4 | |
−21°C | |
borate 1-, tetrahydro-, lithium, unii-8l87x4s4kp, boryllithium, lithium, boryl, lithium hydroborate, lithotab borohydride, bh4li, lithium 1+ ion boranuide, lithium borohydride 2.0 m in thf, borate 1-, tetrahydro-, lithium 1:1 | |
UUKMSDRCXNLYOO-UHFFFAOYSA-N | |
lithium;boron(1-) | |
20722760 | |
AcroSeal™ Glass bottle | |
0.8900g/mL |
16949-15-8, 109-99-9 | |
BH4Li | |
MFCD00011088 | |
15,5581 | |
Solubility in water: decomposes | |
[Li+].[BH4-] | |
21.78 | |
21.77 | |
0.89 |
Safety and Handling
- Lithium borohydride, 4M (10 wt.%) solution in THF
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only