missing translation for 'onlineSavingsMsg'
Learn More

Lithium Carbonate, Honeywell Fluka™

Purum,≥99%

£60.82 - £5585.13

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
PubChem CID 11125
ChEBI CHEBI:6504
IUPAC Name dilithium;carbonate
SMILES [Li+].[Li+].[O-]C([O-])=O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15651230
View Documents
Honeywell-Fluka
13010-100G
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15671230
View Documents
Honeywell-Fluka
13010-1KG
1 kg
£99.47
1kg
Please to purchase this item. Need a web account? Register with us today!
15681230
View Documents
Honeywell-Fluka
13010-6X1KG
6 x 1 kg
£447.58
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15661230
View Documents
Honeywell-Fluka
13010-100KG
100 kg
£5585.13
100kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

554-13-2
73.89
XGZVUEUWXADBQD-UHFFFAOYSA-L
11125
dilithium;carbonate
CLi2O3
MFCD00011084
lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
CHEBI:6504
[Li+].[Li+].[O-]C([O-])=O

Specifications

554-13-2
Li2CO3
NONH for all modes of transport
lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
[Li+].[Li+].[O-]C([O-])=O
73.89
CHEBI:6504
≥99%
Lithium Carbonate
CLi2O3
MFCD00011084
3999191
XGZVUEUWXADBQD-UHFFFAOYSA-L
dilithium;carbonate
11125
73.89g/mol
purum
Videos
Safety and Handling

Safety and Handling

P301 + P312 + P330-P305 + P351 + P338

H302-H319

missing translation for 'einecsNumber' : 209-062-5

missing translation for 'rtecsNumber' : OJ5800000

SDS
Documents

Documents

Product Certifications