missing translation for 'onlineSavingsMsg'
Learn More

Lithium chloride, 99%, For Analysis, Anhydrous, Thermo Scientific Chemicals

Lithium Chloride, 99%, ClLi, CAS Number-7447-41-8, chlorure de lithium, chlorku litu polish, chlorku litu, lithium chloride licl, lithium chloride, licl, luthium chloride, lithiumchloride, lithiumchlorid, chlorolithium, 500g, carbonate (as Li2CO3): < 0.03% (typically), 98.5% min. (Argentometry)

£30.55 - £1759.76

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
IUPAC Name lithium(1+) chloride
SMILES [Li+].[Cl-]
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10532031
missing translation for 'viewDocuments'
Thermo Scientific Acros
199881000
100 g Plastic bottle
£30.55
100g
Please to purchase this item. Need a web account? Register with us today!
10236830
missing translation for 'viewDocuments'
Thermo Scientific Acros
199885000
500 g Plastic bottle
£130.65
500g
Please to purchase this item. Need a web account? Register with us today!
10491241
missing translation for 'viewDocuments'
Thermo Scientific Acros
199880020
2 kg Plastic bottle
£436.17
2kg
Please to purchase this item. Need a web account? Register with us today!
15244674
missing translation for 'viewDocuments'
Thermo Scientific Acros
199880100
10 kg Plastic drum
£1759.76
10kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7447-41-8
42.39
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
433294
lithium(1+) chloride
ClLi
MFCD00011078
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
CHEBI:48607
[Li+].[Cl-]

Specifications

1382.0°C
7447-41-8
Crystalline Granules
ClLi
MFCD00011078
15,5584
Solubility in water: 832g/L (20°C). Other solubilities: soluble in alcohols, ether, pyridine,, nitrobenzene and acetone
[Li+].[Cl-]
42.39
CHEBI:48607
99%
0.5% max. (400°C, 4 hrs)
2.0600g/mL
605.0°C
White
98.5% min. (Argentometry)
LiCl
01,609; 02,246; 04,298; 05,677; 12,277; 16,194
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
lithium(1+) chloride
433294
42.38
Analytical
2.06
Lithium chloride, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 231-212-3

missing translation for 'rtecsNumber' : OJ5950000

missing translation for 'tsca' : TSCA

Product Identifier
  • Lithium chloride, anhydrous

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.