missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- ≥95% for general laboratory use.

Specifications
Specifications
Quantity | 100 g |
Linear Formula | LiNO3 |
UN Number | 2722 |
Solubility Information | Soluble in water |
Formula Weight | 68.95g/mol |
Grade | ReagentPlus™ |
Chemical Name or Material | Lithium Nitrate |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lithium Nitrate, Honeywell >
Spot an opportunity for improvement?Share a Content Correction