Learn More
Lithium sulfate, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals
£144.00 - £1507.00
Chemical Identifiers
CAS | 10377-48-7 |
---|---|
Molecular Formula | Li2O4S |
Molecular Weight (g/mol) | 109.94 |
MDL Number | MFCD00011086 |
InChI Key | INHCSSUBVCNVSK-UHFFFAOYSA-L |
Synonym | lithium sulfate, lithium sulphate, dilithium sulfate, sulfuric acid, dilithium salt, lithiophor, li2so4, dilthium sulfate, sulfuric acid, lithium salt 1:2, unii-919xa137jk, lithium sulfate, anhydrous |
PubChem CID | 66320 |
ChEBI | CHEBI:53474 |
SMILES | [Li+].[Li+].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11418500
|
Thermo Scientific Alfa Aesar
044352.14 |
25 g |
£144.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11408500
|
Thermo Scientific Alfa Aesar
044352.22 |
100 g |
£437.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11428500
|
Thermo Scientific Alfa Aesar
044352.36 |
500 g | |||||||
Description
Lithium sulfate, anhydrous is used to treat bipolar disorder, solar panels and the potential for a new class of battery, and used as a catalyst for the elimination reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10377-48-7 | |
109.94 | |
INHCSSUBVCNVSK-UHFFFAOYSA-L | |
66320 | |
[Li+].[Li+].[O-]S([O-])(=O)=O |
Li2O4S | |
MFCD00011086 | |
lithium sulfate, lithium sulphate, dilithium sulfate, sulfuric acid, dilithium salt, lithiophor, li2so4, dilthium sulfate, sulfuric acid, lithium salt 1:2, unii-919xa137jk, lithium sulfate, anhydrous | |
CHEBI:53474 |
Specifications
845°C | |
White | |
Li2O4S | |
14,5541 | |
Soluble in water(345g/L). Insoluble in absolute ethanol,acetone and pyridine. | |
[Li+].[Li+].[O-]S([O-])(=O)=O | |
109.94 | |
CHEBI:53474 | |
99.99% | |
Hygroscopic | |
(metals basis) |
10377-48-7 | |
Coarse Powder | |
MFCD00011086 | |
lithium sulfate, lithium sulphate, dilithium sulfate, sulfuric acid, dilithium salt, lithiophor, li2so4, dilthium sulfate, sulfuric acid, lithium salt 1:2, unii-919xa137jk, lithium sulfate, anhydrous | |
INHCSSUBVCNVSK-UHFFFAOYSA-L | |
dilithium(1+) sulfate | |
66320 | |
109.94 | |
Odorless | |
2.22 g/mL | |
Lithium sulfate, Anhydrous |
Safety and Handling
GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
H302-H319
missing translation for 'einecsNumber' : 233-820-4
missing translation for 'rtecsNumber' : OJ6419000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only