missing translation for 'onlineSavingsMsg'
Learn More
Lithium tetrakis(pentafluorophenyl)borate ethyl etherate, Thermo Scientific Chemicals Product Code.: 15540574

Lithium tetrakis(pentafluorophenyl)borate ethyl etherate, Thermo Scientific Chemicals

Product Code. 15540574
100 mg
Click to view available options
Quantity:
100 mg
1 g
This item is not returnable. View return policy

Product Code. 15540574

Brand: Thermo Scientific Acros 458531000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 371162-53-7
Molecular Formula 2·5 C4H10O
Molecular Weight (g/mol) 871.28
InChI Key KPLZKJQZPFREPG-UHFFFAOYSA-N
Synonym lithium tetrakis pentafluorophenyl borate ethyl etherate, lithium tetrakis pentafluorophenyl borate, tetrakis pentafluorophenyl boron lithium ethyl etherate, lithium tetrakis pentafluorophenyl borate-ethyl ether complex, lithium tetrakis pentafluorophenyl borate ethyl etherate 2:2:5, lithium tetrakis pentafluorophenyl borate diethylether complex 1:2.5Show MoreShow Less
PubChem CID 23701484
IUPAC Name lithium;ethoxyethane;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
SMILES [Li+].[B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.CCOCC

Specifications

Melting Point 117.0°C to 122.0°C
Quantity 100 mg
Formula Weight 871.28
Chemical Name or Material Lithium tetrakis(pentafluorophenyl)borate ethyl etherate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466364 Certificate TypeCertificate of Analysis Date03/10/2024 Catalog Number458530010,458531000
Lot NumberA0462245 Certificate TypeCertificate of Analysis Date03/05/2024 Catalog Number458530010,458531000
Lot NumberA0460750 Certificate TypeCertificate of Analysis Date31/03/2024 Catalog Number458530010,458531000
Lot NumberA0450238 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number458530010,458531000
Lot NumberA0396931 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number458530010,458531000
compliance-icons
Product Identifier
  • Lithium tetrakis(pentafluorophenyl)borate ethyl etherate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Lithium tetrakis(pentafluorophenyl)borate ethyl etherate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lithium tetrakis(pentafluorophenyl)borate ethyl etherate, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.