Learn More
Lithium tri-tert-butoxyaluminum hydride, nominally 1M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
£42.20 - £127.00
Chemical Identifiers
CAS | 17476-04-9 |
---|---|
Molecular Formula | C12H28AlLiO3 |
MDL Number | MFCD00011532 |
Synonym | Lithium hydridotris(tert-butoxy)aluminate |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11319488
|
Thermo Scientific Alfa Aesar
043324.AB |
10 mL |
£42.20
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11339488
|
Thermo Scientific Alfa Aesar
043324.AD |
50 mL | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Chemical Identifiers
17476-04-9 | |
C12H28AlLiO3 | |
MFCD00011532 | |
Lithium hydridotris(tert-butoxy)aluminate |
Specifications
17476-04-9 | |
0.904 g/mL | |
ChemSeal™ bottles | |
Li[(CH3)3CO]3AlH | |
UN3399 | |
Lithium hydridotris(tert-butoxy)aluminate | |
254.28 | |
Lithium tri-tert-butoxyaluminum hydride |
Colorless | |
−17°C (1°F) | |
C12H28AlLiO3 | |
MFCD00011532 | |
Moisture sensitive | |
It is soluble in organic solvents at low temperatures | |
Liquid |
Safety and Handling
GHS H Statement
H225-H260-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P330-P332+P313-P335+P334
H225-H260-H302-H315-H318-H335-H336-H351
missing translation for 'dotInformation' : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Packaged under Argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only