Learn More
Brannan™ LO-tox™ Lab Thermometer with 3-points Traceable Calibration
LO-tox™ lab thermometer complete with calibration certificate.
Brand:  Brannan™ 44/809/8 + 49/101/0
 View more versions of this product
Product Code. 17823078
  • £121.00 / Each
Explore more special offers
Description
Description
  • LO-tox™ is an organic filling specially chosen to achieve a wide temperature span and provide the lowest practical levels of hazard
  • Safe non-mercury filling
  • Supplied with calibration certificate
  • Non-roll end cap
  • Supplied as standard in protective individual, resealable telescopic tube
  • Made in UK
Specifications
Specifications
0°C, 20°C, 45°C
Total Immersion
Glass
122°F
-31°F
-31°F to +122°F
± 1.0°C at 35°C
1.2 cm
ISO 17025 UKAS
30.5 cm
Blue Spirit
+50°C
-35°C
-35°C to +50°C
0.5°C