missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brannan™ LO-tox™ Lab Thermometer with 3-points Traceable Calibration
LO-tox™ lab thermometer complete with calibration certificate.
Brand: Brannan™ 44/811/8 + 49/101/0
This item is not returnable.
View return policy
Description
- LO-tox™ is an organic filling specially chosen to achieve a wide temperature span and provide the lowest practical levels of hazard
- Safe non-mercury filling
- Supplied with calibration certificate
- Non-roll end cap
- Supplied as standard in protective individual, resealable telescopic tube
- Made in UK
Specifications
0°C, 50°C, 90°C | |
Total Immersion | |
Glass | |
302°F | |
-4°F | |
-4°F to +302°F | |
± 1.5°C at 90°C | |
1.2 cm |
UK Traceable to National Standards ISO 9001 | |
30.5 cm | |
Blue Spirit | |
+150°C | |
-20°C | |
-20°C to +150°C | |
1°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction