missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ IsoGel™ IEF Plate Accessories

Accessories for precast agarose IEF plates. Blotting paper and accessory packs.

Brand:  Lonza™ 56010

 View more versions of this product

Product Code. 11587985

  • £529.00 / Each
Estimated Shipment: 22-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Blotting paper is for use with precast agarose IEF plates
  • Accessory pack includes masks, 100mm and 125mm wicks and blotting paper
  • Accessory bulk pack includes 125mm wicks and blotting paper
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Electrophoresis Accessory Bulk Pack
Dried
18° to 26°C
Isoelectric Focusing
Isoelectric focusing, Antibody separation and analysis
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers