missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ IsoGel™ IEF Plate Accessories
Accessories for precast agarose IEF plates. Blotting paper and accessory packs.
£145.00 - £497.00
Specifications
Separation Type | Isoelectric Focusing |
---|---|
For Use With (Application) | Isoelectric focusing, Antibody separation and analysis |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
11597985
|
Lonza™
56014 |
Accessory Pack |
£145.00
Each |
Estimated Shipment: 24-07-2024
Log in to see stock. |
|||||
11587985
|
Lonza™
56010 |
Electrophoresis Accessory Bulk Pack |
£497.00
Each |
Estimated Shipment: 24-07-2024
Log in to see stock. |
|||||
Description
- Blotting paper is for use with precast agarose IEF plates
- Accessory pack includes masks, 100mm and 125mm wicks and blotting paper
- Accessory bulk pack includes 125mm wicks and blotting paper
Specifications
Isoelectric Focusing | |
Isoelectric focusing, Antibody separation and analysis |