Learn More
SeaKem™ GTG™ Agarose, Lonza™
Specifically designed for preparative DNA electrophoresis. Lonza™ SeaKem™ GTG™ Agarose is a standard gelling temperature, high gel strength agarose that resolves DNA fragments greater than 1,000 bp.
Brand:  Lonza 50070
 View more versions of this product
Product Code. 11530727
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • DNA recovered from SeaKem™ GTG™ Agarose gels may be reliably digested and ligated
  • Extensively performance tested to ensure complete compatibility with routine molecular biology techniques
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SeaKem GTG Agarose
125 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SeaKem™ GTG™ Agarose, Lonza™ > 125g