Learn More
SeaKem™ GTG™ Agarose, Lonza™
Specifically designed for preparative DNA electrophoresis. Lonza™ SeaKem™ GTG™ Agarose is a standard gelling temperature, high gel strength agarose that resolves DNA fragments greater than 1,000 bp.
£215.00 - £2275.00
Specifications
Chemical Name or Material SeaKem GTG Agarose
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11540727
View Documents
Lonza
50071
25 g
£215.00
25g
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
11530727
View Documents
Lonza
50070
125 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11550727
View Documents
Lonza
50074
500 g
£2275.00
500g
Estimated Shipment: 29-05-2025
to see stock.
Add to basket
Description
Description
  • DNA recovered from SeaKem™ GTG™ Agarose gels may be reliably digested and ligated
  • Extensively performance tested to ensure complete compatibility with routine molecular biology techniques
Specifications
Specifications
SeaKem GTG Agarose
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
SeaKem™ GTG™ Agarose, Lonza™