missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Optical Glass Colorimetry Cells

Made from optical glass to the highest standards.

Brand:  Lovibond™ 606130

Code : 04

Additional Details : Weight : 0.09000kg

 View more versions of this product

Product Code. 10436962

  • £109.00 / Each
Estimated Shipment: 16-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Made from optical glass to the highest standards.

Precision fused cells in a range of dimensions and path lengths. Note: For instruments equipped with a heater unit and whenever cells are subjected to thermal shock, it is recommended that borosilicate cells be used.

  • Series: W600 Optical
  • 21 x 40mm cells
  • Use only with nonheated samples (up to 40°C)

Specifications

Specifications

Colorimetry Cell
Model Fs, PFXi-195, 880, 950 and 995 instruments with non-heated Samples (up to 40°C)
Optical Glass
10.79 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers