Learn More
Lovibond™ Replacement Reagents for CHECKIT™ Comparator: Copper No. 2 Tablets
Brand: Lovibond™ 513560BT
Specifications
Yellow | |
5.7 | |
Water analysis | |
0.05 to 5, 0 to 1mg/L | |
Copper No. 2 Tablet |
20 to 25% Boric acid, 60 to 70% Non-hazardous component, 5 to 10% Calcium Thioglycollate Trihydrate, 2.5% Adipic acid | |
Tablet | |
Colorimeter and spectrophotometer | |
Biquinoline |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- copper No. 2 tablets
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P363-Wash contaminated clothing before reuse.
Supplemental information
- MIXTURE LIST-Contains : calcium thioglycolate trihydrate, adipicacid
Your input is important to us. Please complete this form to provide feedback related to the content on this product.