Learn More
Thermo Scientific™ Low DNA Binding Snap Cap Microcentrifuge Tube
GREENER_CHOICE
Microcentrifuge Tubes ensure optimal sample integrity with minimal DNA loss. These tubes provide secure closure and reliable performance, perfect for sensitive applications in molecular biology.
Brand:  Thermo Scientific™ 3405-DLB
 View more versions of this product
Product Code. 30130205
  • £17.85 / Pack of 500
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
The importance of low DNA binding microcentrifuge tubes cannot be overstated. These tubes are specifically engineered to minimize the adhesion of DNA to the tube walls, which is crucial in preserving the quantity and quality of DNA samples during various laboratory procedures.
Ensuring that maximum DNA recovery is achieved and reducing the risk of sample loss and contamination. Particularly vital in applications such as PCR, sequencing, and cloning, where even small losses can significantly impact the results.
Additionally, the assurance that these tubes are free from PCR inhibitors, UV leachables, and human DNA further enhances the reliability of experimental outcomes by preventing any potential interference with sensitive assays.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
Polypropylene Resin
Non-sterile
Snap Cap
PCR Ready
Microcentrifuge Tube
Natural
2 mL
Yes
Cylindrical
Conical
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Brochures
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Low DNA Binding Snap Cap Microcentrifuge Tube >