Learn More
Thermo Scientific™ Low DNA Binding Snap Cap Microcentrifuge Tube

Microcentrifuge Tubes ensure optimal sample integrity with minimal DNA loss. These tubes provide secure closure and reliable performance, perfect for sensitive applications in molecular biology.
£16.80 - £34.40
Specifications
Autoclavable | Autoclavable |
---|---|
Color | Natural |
Material | Polypropylene Resin |
Sterility | Non-sterile |
Graduated | Yes |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
30130204
![]() |
Thermo Scientific™
3403-DLB |
0.6 mL |
£34.40
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
||||||
30130203
![]() |
Thermo Scientific™
3404-DLB |
1.5 mL |
£16.80
Pack of 500 |
In Stock
Log in to see stock. ![]() |
||||||
30130205
![]() |
Thermo Scientific™
3405-DLB |
2 mL |
£17.85
Pack of 500 |
In Stock
Log in to see stock. ![]() |
||||||
Description
The importance of low DNA binding microcentrifuge tubes cannot be overstated. These tubes are specifically engineered to minimize the adhesion of DNA to the tube walls, which is crucial in preserving the quantity and quality of DNA samples during various laboratory procedures.
Ensuring that maximum DNA recovery is achieved and reducing the risk of sample loss and contamination. Particularly vital in applications such as PCR, sequencing, and cloning, where even small losses can significantly impact the results. Additionally, the assurance that these tubes are free from PCR inhibitors, UV leachables, and human DNA further enhances the reliability of experimental outcomes by preventing any potential interference with sensitive assays.Specifications
Autoclavable | |
Polypropylene Resin | |
Yes | |
Cylindrical | |
Conical |
Natural | |
Non-sterile | |
Snap Cap | |
PCR Ready | |
Microcentrifuge Tube |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.