Learn More
Porcelaines Avignon™ Low Form Porcelain Crucibles
Heat and cool substances in the laboratory using Porcelaines Avignon™ Low Form Porcelain Crucibles. They are available in various capacities.
£73.90
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11748473
|
Porcelaines Avignon™
11306 |
40 mL | N/A | ||||||
Description
- Low walls
- Withstand high temperatures and rapid temperature changes
- Porcelain
Specifications
Porcelain | |
Crucible |