missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ LPS from E. Coli, TLR4 ligand

Applications: Functional

Brand:  Novus Biologicals™ NBP2-25295-1.0mg

Additional Details : Weight : 0.01000kg

Product Code. 18656528

  • £192.22 / 1mg
Estimated Shipment: 17-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Lipopolysaccharides from Escherichia coli 0111:B4 Gamma-irradiated, BioXtra, suitable for cell culture Dilution: Functional Purity: for ion exchange chromatography Reconstitute with 1 ml sterile balanced salt solution or tissue culture medium to the vial (1 mg) and gently swirl until the powder dissolves. Reconstituted product may be further diluted to desired working concentrations.
Specifications

Specifications

LYOPH
1 mg
LPS from E. Coli, TLR4 Ligand
Store at -20C. Avoid freeze-thaw cycles.
Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular-patterns (PAMPs). Use in functional reported in scientific literature (PMID 26121241).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers