Learn More
GeneBLAzer™ LTB4R-Gα15-NFAT-bla CHO-K1 Cells
GeneBLAzer™ LTB4R-Gα15-NFAT-bla CHO-K1 Cells contain the human Leukotriene B4 Receptor.
Brand:  GeneBLAzer™ K1748
Code : NEW
Additional Details : Weight : 0.01000kg
Product Code. 10328763
  • £10780.00 / Each
Estimated Shipment: 05-08-2024
to see stock.
Add to basket
Description
Description
  • Gene Symbol: LTB4R
  • Accession Number: NM_181657
  • Population Status: Single Cell Clone Origin
  • CellSensor™ NFAT-bla Freestyle HEK 293F cells (Cat. no. K1725) contain a beta-lactamase reporter gene under control of the NFAT Response Element.
  • GeneBLAzer™ LTB4R-Gα15-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of LTB4. In addition, GeneBLAzer™ LTB4R-Gα15-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions.
Specifications
Specifications
Should be stored in liquid nitrogen immediately upon receipt
GPCRs
2 x 106 Cells
Fluorescent
LTB4R-Gα15-NFAT-bla CHO-K1 cells, LTB4R-Gα15-NFAT-bla CHO-K1 Assay Protocol, Certificate of Analysis
Dividing Cells