missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Luc-Screen™ Extended-Glow Luciferase Reporter Gene Assay System

Thermo Scientific Luc-Screen Extended-Glow Luciferase Reporter Gene Assay System with extended-glow light emission is designed for sensitive detection of firefly luciferase reporter enzyme in microplate cell cultures. A linear signal is obtained with the

Brand:  Invitrogen™ T1035

 View more versions of this product

Product Code. 12634295

  • £221.00 / Each
Estimated Shipment: 05-12-2024
to see stock.
Explore more special offers
Add to basket

Includes: Two reaction buffers
This item is not returnable. View return policy

Description

Description

  • Homogeneous assay format allows detection of firefly luciferase in presence of normal culture media without removal of media and without an additional cell lysis step, providing easiest and most streamlined assay procedure possible
  • Glow assay kinetics provides a “window” during which measurements may be performed, facilitating HTS applications where assay automation is used
  • Wide dynamic range of firefly luciferase assay lets the user measure enzyme level accurately from femtogram to nanogram range
  • Assay sensitivity is 100- to 1,000-fold better than with either the isotopic/non-isotopic assays for chloramphenicol acetyl transferase (CAT) or the colorimetric/fluorescent assays for β-galactosidase, providing greater sensitivity than competing assay technologies
  • Highly sensitive assay with a wide dynamic range permits detection of high and low levels of reporter without performing numerous sample dilutions
  • Non-radioactive reporter gene assay kit eliminates radioisotopes
  • Assay can be completed in about one hour, providing fast assay turnaround
Sensitive
Luciferase is an ideal reporter because of the high sensitivity of detection and the absence of endogenous luciferase activity in mammalian cells. The Luc-Screen assay conveniently couples in-well cell lysis in the presence of culture medium with a high sensitivity assay that exhibits extended-glow light emission kinetics. Light signal can be measured between 10 minutes and several hours after adding assay reagents, providing great flexibility in the time between reagent addition and measurement.
 
High Throughput
The Luc-Screen system is designed for maximum assay flexibility in a high throughput format and can be used in luminometers without automatic injectors. The Luc-Screen system is formulated to provide a convenient, easy-to-use luciferase assay that is optimized for use in high-throughput screening. The high sensitivity of the Luc-Screen assay is accompanied by a wide dynamic range.
 
Specifications
  • Assay Type: Luciferase Assay
  • Compatible Cells: Mammalian Cells
  • Detection Method: Chemiluminescent
  • For Use With Equipment: Luminometer (Microplate)
  • Form: Liquid
  • Format: 96-well plate
  • Readout: End Point
  • Sample Type (Specific): Cell Cultures
  • Substrate: Luciferin
  • Substrate Properties: Chemical Substrate
  • Substrate Type: Luciferase Substrate
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Luciferase Reporter Gene Assay System
Luminometer (Microplate)
This kit contains two reaction buffers.
Store the product at -20°C.
High-throughput Compatible
Cell Cultures
Luciferase Substrate
Luciferase
Enzyme Assay
Liquid
Chemiluminescence
Luciferase Assay
Mammalian Cells
End Point
Luciferin
Chemical Substrate
Luc-Screen™, NovaBright™
200 Assays
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.