Learn More
Emma Safety Footwear Lukas Safety Shoes
Includes a steel toe cap, a polyurethane outer toe cap and an easy-twist sole that provides a sticky grip. Available in a variety of widths.
Marca: Emma Safety Footwear MM73054838
Detalles adicionales : Weight : 1.59800kg
Descripción
- S3-Safety shoe with sole plate for wet work
- Full grain leather
- GripForce™ Easy Twist
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Especificaciones
EN ISO 20345:2011 | |
Black | |
Hydro-Tec™ Sanitized Silver | |
38 |
Laces | |
Lukas | |
Leather | |
Steel |