missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11343467
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Lutetium(III) chloride, anhydrous is used as a sputtering target in the preparation of bubble memory device. It is widely used in electronic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10099-66-8
MDL Number MFCD00011099

Specifications

Melting Point 905°C (sublimation)
Quantity 1 g
Merck Index 14,5615
Solubility Information Soluble in water.
Formula Weight 281.33
Percent Purity 99.9%
Sensitivity Hygroscopic
Density 3.98 g/mL
Assay (REO)
Chemical Name or Material Lutetium(III) chloride, Anhydrous

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.