missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) nitrate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11312518
Click to view available options
Quantity:
1 g
5 g
25 g

Lutetium(III) nitrate hydrate is used in ceramics, glass, phosphors and in lasers. It finds application as a catalyst in petroleum cracking and to prepare laser crystal. Further, it is used in hydrogenation, alkylation and polymerization reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100641-16-5
Molecular Formula LuN3O9
Molecular Weight (g/mol) 360.98
MDL Number MFCD00150421
InChI Key APRNQTOXCXOSHO-UHFFFAOYSA-N
Synonym lutetium iii nitrate hydrate, lutetium nitrate hydrate, acmc-1buji, lu.3no3.h2o, lutetium nitrate-water 1/3/1, lutetium 3+ hydrate trinitrate
PubChem CID 16212852
SMILES [Lu+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color White
Physical Form Crystalline Aggregates
Quantity 5 g
Linear Formula Lu(NO3)3·xH2O
UN Number UN1477
Solubility Information Soluble in water. Slightly soluble in strong mineral acids.
IUPAC Name lutetium(3+) trinitrate
Formula Weight 360.98 (Anhydrous)
Percent Purity 99.9%
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Lutetium(III) nitrate hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'