Learn More
Welch™ LVS 101 Z Laboratory Vacuum System
Modular manual and automatic control chemically resistant PTFE construction
Brand:  Welch™ 115027
Product Code. 11849081
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Modular manual and automatic control chemically resistant PTFE construction 230V 50Hz flow rate 17L/min vacuum mbar <8 Ilmvac
TRUSTED_SUSTAINABILITY
Specifications
Specifications
230 V 50/60 Hz
LVS 101 Z
445 mm
Laboratory Vacuum System
230 V
310 mm
50/60 Hz
360 mm
<8 mbar
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Welch™ LVS 101 Z Laboratory Vacuum System >